ContactUsLinkComponent
Refill therm. for sling psychrometer, solid, -10+60:0,2C, milk glass scale, capillary tube specially coated inside, with non-wetti ng blue special liquid, 280x8mm, null
Catalog # AMARK175228-CB
Supplier:  Amarell
Refill therm. for sling psychrometer, solid, -10+60:0,2C, milk glass scale, capillary tube specially coated inside, with non-wetti ng blue special liquid, 280x8mm, null
Catalog # AMARK175228-CB
Supplier:  Amarell
Frequently Bought Together