ContactUsLinkComponent
Stirring therm., enclosed scale, -10+150:1C, 260x7-8mm, cap. prismatic, colourless, red special liquid, total immersion, with ring at top 1 * 1 items
Catalog # AMARG11914-FL
Supplier:  Amarell
Stirring therm., enclosed scale, -10+150:1C, 260x7-8mm, cap. prismatic, colourless, red special liquid, total immersion, with ring at top 1 * 1 items
Catalog # AMARG11914-FL
Supplier:  Amarell
Frequently Bought Together