ContactUsLinkComponent
Therm. for demonstration purposes, -50+150:1C, milk-glass scale, red special liquid, upper part c. 500x23mm, stem of 100x9mm = imme rsion, top finish: cork and metal cap 1 * 1 items
Catalog # AMARG11730-FL
Supplier:  Amarell
Therm. for demonstration purposes, -50+150:1C, milk-glass scale, red special liquid, upper part c. 500x23mm, stem of 100x9mm = imme rsion, top finish: cork and metal cap 1 * 1 items
Catalog # AMARG11730-FL
Supplier:  Amarell
Frequently Bought Together