ContactUsLinkComponent
Precision therm., low temp., DIN 12778, -50+50:1C, solid stem, white backed, toluene filled, 300x6-7mm, with ring at top, suitable for government verification 1 * 1 items
Catalog # AMARL33090-TOL
Supplier:  Amarell
Precision therm., low temp., DIN 12778, -50+50:1C, solid stem, white backed, toluene filled, 300x6-7mm, with ring at top, suitable for government verification 1 * 1 items
Catalog # AMARL33090-TOL
Supplier:  Amarell
Frequently Bought Together