Thermometer, solid stem, similar to ASTM 78 F, white backed, 295+315:0,5F, capillary tube specially coated inside, with non-wettin g blue special liquid, total immersion, max.280x6-7mm, durable pigment, null
Catalog # AMARA301090-CB
Supplier: Amarell
Frequently Bought Together