ContactUsLinkComponent
Therm. with standard ground joint 10/30, solid stem, white backed, built-in length 105mm, -10+360:1C, upper part 300x6-7mm, eco-fri endly, green liquid, biodegradable 1 * 1 items
Catalog # AMARD264106-EF
Supplier:  Amarell
Therm. with standard ground joint 10/30, solid stem, white backed, built-in length 105mm, -10+360:1C, upper part 300x6-7mm, eco-fri endly, green liquid, biodegradable 1 * 1 items
Catalog # AMARD264106-EF
Supplier:  Amarell
Frequently Bought Together