ContactUsLinkComponent
Precision therm., high temp., similiar DIN, +200+610:1C, solid stem, frosted back, total immersion, gallium filled, 450mm long, dia meter 6-7mm, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARL29604-GA
Supplier:  Amarell
Precision therm., high temp., similiar DIN, +200+610:1C, solid stem, frosted back, total immersion, gallium filled, 450mm long, dia meter 6-7mm, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARL29604-GA
Supplier:  Amarell
Frequently Bought Together