Precision thermometer, similar DIN, 0+100:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 550x6-7mm, durable pigment, null
Catalog # AMARL27382-CB
Supplier: Amarell
Frequently Bought Together