ContactUsLinkComponent
Thermometer similar to AFNOR, STL/0,5/190/310, +190+310:0,5C, solid stem, white backed, gallium filled, 375mm long, total immersion , durable pigment, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARA411074-GA
Supplier:  Amarell
Thermometer similar to AFNOR, STL/0,5/190/310, +190+310:0,5C, solid stem, white backed, gallium filled, 375mm long, total immersion , durable pigment, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARA411074-GA
Supplier:  Amarell
Frequently Bought Together