ContactUsLinkComponent
Thermometer similar to AFNOR, STL/0,2/-55/5, -55+5:0,2C, solid stem, white backed, toluene filled, 375mm long, total immersion, sui table for government calibration 1 * 1 items
Catalog # AMARA411032-TOL
Supplier:  Amarell
Thermometer similar to AFNOR, STL/0,2/-55/5, -55+5:0,2C, solid stem, white backed, toluene filled, 375mm long, total immersion, sui table for government calibration 1 * 1 items
Catalog # AMARA411032-TOL
Supplier:  Amarell
Frequently Bought Together