ContactUsLinkComponent
Distillation therm., similar to DIN 12779, +200+360:1C, enclosed scale, cap. prismatic, colourless, gallium filled, total immersion , length 210mm, pin-top finish with ring 1 * 1 items
Catalog # AMARD259614-GA
Supplier:  Amarell
Distillation therm., similar to DIN 12779, +200+360:1C, enclosed scale, cap. prismatic, colourless, gallium filled, total immersion , length 210mm, pin-top finish with ring 1 * 1 items
Catalog # AMARD259614-GA
Supplier:  Amarell
Frequently Bought Together