Beer hydrometer, 0,9974-1,0036:0,0002g/cm , with therm. +13+26:0,2C with red or blue special liquid, with correction scale, reading above, ref. temp. 20C, 415mm long 1 * 1 items
Catalog # AMARH847500-FL
Supplier: Amarell
Frequently Bought Together