ContactUsLinkComponent
Alcoholometer, similar to EUROPA class III, 25-30:0,1%vol., similar DIN 12803, with therm. +5+25:0,2C with red or blue speclial liq uid, 330mm long, suitable for government verification 1 * 1 items
Catalog # AMARH848230-FL
Supplier:  Amarell
Alcoholometer, similar to EUROPA class III, 25-30:0,1%vol., similar DIN 12803, with therm. +5+25:0,2C with red or blue speclial liq uid, 330mm long, suitable for government verification 1 * 1 items
Catalog # AMARH848230-FL
Supplier:  Amarell
Frequently Bought Together