ContactUsLinkComponent
Thermomometer similar to AFNOR, STL 0,1/40/65, +40+65:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 375mm long, total immersion, with ice point scale, null
Catalog # AMARA411006-CB
Supplier:  Amarell
Thermomometer similar to AFNOR, STL 0,1/40/65, +40+65:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 375mm long, total immersion, with ice point scale, null
Catalog # AMARA411006-CB
Supplier:  Amarell
Frequently Bought Together