ContactUsLinkComponent
Thermometer similar to AFNOR, STL/0,1/20/45, +20+45:0,1C, solid stem, white backed, capillary tube specially coated inside, with n on-wetting blue special liquid, 375mm long, total immersion, durable pigment, with ice point scale, null
Catalog # AMARA411004-CB
Supplier:  Amarell
Thermometer similar to AFNOR, STL/0,1/20/45, +20+45:0,1C, solid stem, white backed, capillary tube specially coated inside, with n on-wetting blue special liquid, 375mm long, total immersion, durable pigment, with ice point scale, null
Catalog # AMARA411004-CB
Supplier:  Amarell
Frequently Bought Together