ContactUsLinkComponent
Thermometer similar to BS 593, F200C/100, 148+202:0,2C, solid stem, white backed, gallium filled, 395mm long, 100mm immersion, dura ble pigment, suitable for government verification 1 * 1 items
Catalog # AMARA403392-GA
Supplier:  Amarell
Thermometer similar to BS 593, F200C/100, 148+202:0,2C, solid stem, white backed, gallium filled, 395mm long, 100mm immersion, dura ble pigment, suitable for government verification 1 * 1 items
Catalog # AMARA403392-GA
Supplier:  Amarell
Frequently Bought Together