ContactUsLinkComponent
Thermometer similar to BS 593, B260C/100, 200+260:0,2C, solid stem, white backed, gallium filled, 400mm long, 100mm immersion, dura ble pigment, with ice point scale, suitable for government calibration 1 * 1 items
Catalog # AMARA403262-GA
Supplier:  Amarell
Thermometer similar to BS 593, B260C/100, 200+260:0,2C, solid stem, white backed, gallium filled, 400mm long, 100mm immersion, dura ble pigment, with ice point scale, suitable for government calibration 1 * 1 items
Catalog # AMARA403262-GA
Supplier:  Amarell
Frequently Bought Together