Pocket saccharimeter, 0-7:0,1%mas., with therm. +10+25:0,2C with red or blue special liquid, with correction scale, reading above, ref. temp. 20C, 275mm long, suitable for government calibration 1 * 1 items
Catalog # AMARH847480-FL
Supplier: Amarell
Frequently Bought Together