ContactUsLinkComponent
Sweet wort hydrometer, -1+2:0,05%mas, with therm. +5+30:0,5C with red or blue special liquid, with correction, reading below, ref. temp. 20C, 400mm long 1 * 1 items
  AMARH847348-FL
 :  Amarell
Sweet wort hydrometer, -1+2:0,05%mas, with therm. +5+30:0,5C with red or blue special liquid, with correction, reading below, ref. temp. 20C, 400mm long 1 * 1 items
  AMARH847348-FL
 :  Amarell
Frequently Bought Together