Thermometer similar to BS 593, A40C/100, 0+40:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-wet ting blue special liquid, 400mm long, 100mm immersion, durable pigment, null
Catalog # AMARA403244-CB
Supplier: Amarell
Frequently Bought Together