ContactUsLinkComponent
Thermometer similar to BS 593, B60C/total, solid stem, white backed, -20+60:0,2C, capillary tube specially coated inside, with non -wetting blue special liquid, 400mm long, total immersion, durable pigment, null
Catalog # AMARA403212-CB
Supplier:  Amarell
Thermometer similar to BS 593, B60C/total, solid stem, white backed, -20+60:0,2C, capillary tube specially coated inside, with non -wetting blue special liquid, 400mm long, total immersion, durable pigment, null
Catalog # AMARA403212-CB
Supplier:  Amarell
Frequently Bought Together