Thermometer similar to BS 1365, SB195C/total, 185+195:0,1C, solid stem, white backed, gallium filled, 235mm long, total immersion, durable pigment, suitable for government calibration 1 * 1 items
Catalog # AMARA402580-GA
Supplier: Amarell
Frequently Bought Together