Thermometer similar to BS 1365, SB145C/total, 135+145:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402570-CB
Supplier: Amarell
Frequently Bought Together