ContactUsLinkComponent
Thermometer similar to BS 1365, SB115C/total, 105+115:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402564-CB
Supplier:  Amarell
Thermometer similar to BS 1365, SB115C/total, 105+115:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402564-CB
Supplier:  Amarell
Frequently Bought Together