Thermometer similar to BS 1365, BS35C/total, 25+35:0,1C, solid stem, white backed, capillary tube specially coated inside, with no n-wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402548-CB
Supplier: Amarell
Frequently Bought Together