ContactUsLinkComponent
Thermometer similar to BS 1365, SB1C/total, -10+1:0,1C, solid stem, white backed, capillary tube specially coated inside, with non -wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402540-CB
Supplier:  Amarell
Thermometer similar to BS 1365, SB1C/total, -10+1:0,1C, solid stem, white backed, capillary tube specially coated inside, with non -wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402540-CB
Supplier:  Amarell
Frequently Bought Together