ContactUsLinkComponent
Thermometer similar to BS 1365, SA155C/total, 94+155:0,5C, solid stem, white backed, red special liquid, 215mm long, total immersio n, durable pigment, suitable for government calibration 1 * 1 items
  AMARA402504-FL
 :  Amarell
Thermometer similar to BS 1365, SA155C/total, 94+155:0,5C, solid stem, white backed, red special liquid, 215mm long, total immersio n, durable pigment, suitable for government calibration 1 * 1 items
  AMARA402504-FL
 :  Amarell
Frequently Bought Together