ContactUsLinkComponent
Thermometer similar to BS 1704, C/75, 0+60:0,5C, solid stem, white backed, capillary tube specially coated inside, with non-wettin g blue special liquid, 75mm immersion, 300mm long, durable pigment, null
Catalog # AMARA402104-CB
Supplier:  Amarell
Thermometer similar to BS 1704, C/75, 0+60:0,5C, solid stem, white backed, capillary tube specially coated inside, with non-wettin g blue special liquid, 75mm immersion, 300mm long, durable pigment, null
Catalog # AMARA402104-CB
Supplier:  Amarell
Frequently Bought Together