ContactUsLinkComponent
Thermometer similar to BS 1704, D/total, 0+100:1C, solid stem, white backed, capillary tube specially coated inside, with non-wett ing blue special liquid, total immersion, 300mm long, durable pigment, null
Catalog # AMARA402086-CB
Supplier:  Amarell
Thermometer similar to BS 1704, D/total, 0+100:1C, solid stem, white backed, capillary tube specially coated inside, with non-wett ing blue special liquid, total immersion, 300mm long, durable pigment, null
Catalog # AMARA402086-CB
Supplier:  Amarell
Frequently Bought Together