ContactUsLinkComponent
Hydrometer for liquefied gas, suitable for overpressure of 14bar, with therm. 0+30:1C with red or blue special liquid, 0,550-0,600: 0,001g/cm , ref. temp. 15C, 360mm long 1 * 1 items
Catalog # AMARH838762-FL
Supplier:  Amarell
Hydrometer for liquefied gas, suitable for overpressure of 14bar, with therm. 0+30:1C with red or blue special liquid, 0,550-0,600: 0,001g/cm , ref. temp. 15C, 360mm long 1 * 1 items
Catalog # AMARH838762-FL
Supplier:  Amarell
Frequently Bought Together