ContactUsLinkComponent
Thermometer similar to BS 1900, SR5/80C, 69,5+80,5:0,02C, length 480mm, total immersion, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, with ice point scale, null
Catalog # AMARA401922-CB
Supplier:  Amarell
Thermometer similar to BS 1900, SR5/80C, 69,5+80,5:0,02C, length 480mm, total immersion, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, with ice point scale, null
Catalog # AMARA401922-CB
Supplier:  Amarell
Frequently Bought Together