ContactUsLinkComponent
Thermometer similar to BS 1900, SR5/40C, +29,5+40,5:0,02C, length 480mm, total immersion, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, with ice point scale, null
Catalog # AMARA401914-CB
Supplier:  Amarell
Thermometer similar to BS 1900, SR5/40C, +29,5+40,5:0,02C, length 480mm, total immersion, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, with ice point scale, null
Catalog # AMARA401914-CB
Supplier:  Amarell
Frequently Bought Together