ContactUsLinkComponent
Thermometer similar to BS 1900, SR5/30C, 19,5+30,5:0,02C, length 480mm, total immersion, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, with ice point scale, null
  AMARA401912-CB
 :  Amarell
Thermometer similar to BS 1900, SR5/30C, 19,5+30,5:0,02C, length 480mm, total immersion, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, with ice point scale, null
  AMARA401912-CB
 :  Amarell
Frequently Bought Together