ContactUsLinkComponent
Thermometer similar to BS 1900, SR5/20C, 9,5+20,5:0,02C, length 480mm, total immersion, white backed, capillary tube specially coa ted inside, with non-wetting blue special liquid, with ice point scale, null
Catalog # AMARA401910-CB
Supplier:  Amarell
Thermometer similar to BS 1900, SR5/20C, 9,5+20,5:0,02C, length 480mm, total immersion, white backed, capillary tube specially coa ted inside, with non-wetting blue special liquid, with ice point scale, null
Catalog # AMARA401910-CB
Supplier:  Amarell
Frequently Bought Together