ContactUsLinkComponent
Thermometer similar to S.T.P.T.C. T 24d, solid stem, white backed, 72+82:0,05C, length 400mm, imm. 100mm, capillary tube specially coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400190-CB
Supplier:  Amarell
Thermometer similar to S.T.P.T.C. T 24d, solid stem, white backed, 72+82:0,05C, length 400mm, imm. 100mm, capillary tube specially coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400190-CB
Supplier:  Amarell
Frequently Bought Together