ContactUsLinkComponent
Thermometer similar to S.T.P.T.C. T 9d, solid stem, white backed, 50+210:0,5C, length 430mm, imm. 100mm, red special liquid, durabl e pigment, suitable for government calibration 1 * 1 items
Catalog # AMARA400060-FL
Supplier:  Amarell
Thermometer similar to S.T.P.T.C. T 9d, solid stem, white backed, 50+210:0,5C, length 430mm, imm. 100mm, red special liquid, durabl e pigment, suitable for government calibration 1 * 1 items
Catalog # AMARA400060-FL
Supplier:  Amarell
Frequently Bought Together