ContactUsLinkComponent
Sample Tray G, polypropylene, for Agilent integrated autosampler (I-AS), with 28 positions, for 20 mL PFA vials up to 28 mm od x 58 mm tall, suitable for use in semiconductor applications. Vials should be ordered separately 1 * 1 items
Catalog # HEWLG3160-80066
Supplier:  Agilent
Sample Tray G, polypropylene, for Agilent integrated autosampler (I-AS), with 28 positions, for 20 mL PFA vials up to 28 mm od x 58 mm tall, suitable for use in semiconductor applications. Vials should be ordered separately 1 * 1 items
Catalog # HEWLG3160-80066
Supplier:  Agilent
Frequently Bought Together