ContactUsLinkComponent
Bottle head assembly, for use with the 1260/1290 Infinity II Preperative Pumps G7161A/B, pre-assembled, including tube ESD-PTFE 2000 mm length, bottle cap, and solvent inlet filter 20 µm 1 * 1 items
Catalog # HEWLG7161-60035
Supplier:  Agilent
Bottle head assembly, for use with the 1260/1290 Infinity II Preperative Pumps G7161A/B, pre-assembled, including tube ESD-PTFE 2000 mm length, bottle cap, and solvent inlet filter 20 µm 1 * 1 items
Catalog # HEWLG7161-60035
Supplier:  Agilent
Frequently Bought Together