Length 600mm, diameter 10mm, PTFE-coated stainless steel shaft, U-shaped stirring blade made entirely of PTFE. 1 * 1 items
Catalog # BOHLC384-22
Supplier: Bohlender
Length 600mm, diameter 10mm, PTFE-coated stainless steel shaft, U-shaped stirring blade made entirely of PTFE. 1 * 1 items
Catalog # BOHLC384-22
Supplier: Bohlender
Frequently Bought Together
Accessories for vortex mixers, MS 3
Supplier: IKA
280 x 170 mm Low-Adhesion Mat, pack of 2 for Solaris Shaker Platforms, can be cut to size 1 * 1 items
Supplier: Thermo Fisher Scientific
Thermo Scientific™ Digital Microplate Shaker
Supplier: Thermo Fisher Scientific
Accessories for overhead stirrers RW 47 D
Supplier: IKA
Cole-Parmer® Stuart RS-200 Roller Shakers
Supplier: Cole Parmer
Accessories for vortex mixers Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Accessories for Shakers, SM 30 A and SM 30 A control and SM 30 B and SM 30 B control
Supplier: Edmund Bühler GmbH
Stirrer bearings, PTFE
Supplier: Bohlender
MaxQ™ 8000 Incubating Shakers
Supplier: Thermo Fisher Scientific
PressureSyn Glass Liner 1 * 1 items
Supplier: ASYNT
Grinding balls for planetary ball mills, PM 100, PM 100 CM, PM 200 and PM 400
Supplier: Retsch GmbH
Accessories for heating bath, HBR 4 digital
Supplier: IKA
Accessories for TW alpha plus
Supplier: SI Analytics
Accessories for Ultra-centrifugal mill, ZM 200
Supplier: Retsch GmbH
Grinding balls for MM 200 and MM 400
Supplier: Retsch GmbH
DrySyn® heating blocks
Supplier: ASYNT