ContactUsLinkComponent
EC HPLC column EC 125/4 NUCLEOSIL 100-10 C18 length: 125 mm, ID: 4 mm pack of 1 1 * 1 items
Catalog # MANA720701.40
Supplier:  MACHEREY-NAGEL
EC HPLC column EC 125/4 NUCLEOSIL 100-10 C18 length: 125 mm, ID: 4 mm pack of 1 1 * 1 items
Catalog # MANA720701.40
Supplier:  MACHEREY-NAGEL
Frequently Bought Together