ContactUsLinkComponent
Test Sieve for cereals with stainless steel frame 200x32 mm, stainless steel-perforated plate with slot holes according to ISO 5223 1 * 1 items
Catalog # HAVE206160966
Supplier:  NEXOPART
Test Sieve for cereals with stainless steel frame 200x32 mm, stainless steel-perforated plate with slot holes according to ISO 5223 1 * 1 items
Catalog # HAVE206160966
Supplier:  NEXOPART
Frequently Bought Together