Coumalic acid
Bottle, 5 g
APOSOR22935-5G
: 2-Oxo-2H-pyran-5-carboxylic acid
- :500-05-0
- :C₆H₄O₄
- :MFCD00006644
- :140.10 g/mol
- :218 °C/120mmHg
- :203…205 °C (dec.)
- :Ambient
- Pack type:Bottle
- Pk:5 g