Gö 6983 ≥99%
Bottle, 1 mg
Catalog # APOSBIFK0056-1MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Gö 6983 is a potent, cell-permeable, ATP-competitive inhibitor of protein kinase C (PKC).
- Free base
- Purity: >99%.
Synonyms: 3-(1-(3-(Dimethylamino)propyl)-5-methoxy-1H-indol-3-yl)-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione, 3-[1-[3-(dimethylamino)propyl]-5-methoxy-1H-indol-3-yl]-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione, Go 6983
- CAS Number:133053-19-7
- Formula:C₂₆H₂₆N₄O₃
- MDL Number:MFCD04040031
- Molecular Weight:442.519 g/mol
- Storage Temperature:Freezer
Specifications
Specifications
- Pack type:Bottle
- Pk:1 mg



