ICP Multi-Element Standard Solution 28 elements in 2 % HNO3, 1mg/l 1 * 100 mL
Catalog # ROTH2649.1
Supplier: Roth Carl
ICP Multi-Element Standard Solution 28 elements in 2 % HNO3, 1mg/l 1 * 100 mL
Catalog # ROTH2649.1
Supplier: Roth Carl
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Humidity standards
Supplier: ROTRONIC INSTRUMENTS
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Safety Waste Caps, Electrically Conductive
Supplier: S.C.A.T.
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
pH buffers, colour coded, reference standards, Supelco®
Supplier: Merck