ContactUsLinkComponent
N,N'-Dicyclohexylcarbodiimide (DCC) ?98 %, for peptide synthesis Coupling reagent for peptide synthesis DCC Empirical formula C13H22N2 Molar mass (M) 206,33 g/mol Boiling point (bp) 32-35 °C Solubility 10 mg/l (H2O, 20 °C) ADR 6.1 II • WGK 3 CAS-Nr. 1 * 250 g
Catalog # ROTH4193.2
Supplier:  Roth Carl
N,N'-Dicyclohexylcarbodiimide (DCC) ?98 %, for peptide synthesis Coupling reagent for peptide synthesis DCC Empirical formula C13H22N2 Molar mass (M) 206,33 g/mol Boiling point (bp) 32-35 °C Solubility 10 mg/l (H2O, 20 °C) ADR 6.1 II • WGK 3 CAS-Nr. 1 * 250 g
Catalog # ROTH4193.2
Supplier:  Roth Carl
Frequently Bought Together