ContactUsLinkComponent
Pre-filled Milliflex Cassettes : Aerobic Heterotrophic microorganisms, Tryptic Soy Agar with poly 80 and Lecithin, 48 cassettes/pk 1 * 48 items
Catalog # MILXMXSMTLP48
Supplier:  Merck
Pre-filled Milliflex Cassettes : Aerobic Heterotrophic microorganisms, Tryptic Soy Agar with poly 80 and Lecithin, 48 cassettes/pk 1 * 48 items
Catalog # MILXMXSMTLP48
Supplier:  Merck
Frequently Bought Together