ContactUsLinkComponent
Pressure transmitter for general applications Model S-10 Specifications according to data sheet: PE 81.01 Power supply: DC 10 (14) ... 30 V Wetted parts: stainless steel Case: stainless steel Output signal: 4 ... 20 mA, 2-wire Measuring range: 0 ... 1 * 1 items
Catalog # IBSA7728073
Supplier:  IBS Stock Items
Pressure transmitter for general applications Model S-10 Specifications according to data sheet: PE 81.01 Power supply: DC 10 (14) ... 30 V Wetted parts: stainless steel Case: stainless steel Output signal: 4 ... 20 mA, 2-wire Measuring range: 0 ... 1 * 1 items
Catalog # IBSA7728073
Supplier:  IBS Stock Items
Frequently Bought Together