ContactUsLinkComponent
CrystalBreeder Multiple reactor system 32 reactors 8 temperature zones temperature accuracy, 0.1 C heating/cooling rate, 0-20 C/minute temperature range -15C to 150C* variable magnetic stirring in each reactor including overhead stirring turbidit 1 * 1 items
Catalog # IBSA3002-100-000
Supplier:  IBS Stock Items
CrystalBreeder Multiple reactor system 32 reactors 8 temperature zones temperature accuracy, 0.1 C heating/cooling rate, 0-20 C/minute temperature range -15C to 150C* variable magnetic stirring in each reactor including overhead stirring turbidit 1 * 1 items
Catalog # IBSA3002-100-000
Supplier:  IBS Stock Items
Frequently Bought Together