ContactUsLinkComponent
Silicon tube set included with AVS®/S and AVS®/SK, to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 .., 538 .., 5 47 .., 548 ..and 549 .. to a AVS® 310, 350, 450 and 470 1 * 1 items
Catalog # SCOI285414027
Supplier:  SI Analytics
Silicon tube set included with AVS®/S and AVS®/SK, to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 .., 538 .., 5 47 .., 548 ..and 549 .. to a AVS® 310, 350, 450 and 470 1 * 1 items
Catalog # SCOI285414027
Supplier:  SI Analytics
Frequently Bought Together