ContactUsLinkComponent
384 well cell repellent microplate, black uclear, individually wrapped, sterile with lid 1 * 32 items
Catalog # GREI781976-SIN
Supplier:  Greiner Bio-One
384 well cell repellent microplate, black uclear, individually wrapped, sterile with lid 1 * 32 items
Catalog # GREI781976-SIN
Supplier:  Greiner Bio-One